Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC8 Monoclonal Antibody (1A5B9)
GREENER_CHOICE

Catalog No. FERMA548730
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548730 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548730 Supplier Invitrogen™ Supplier No. MA548730
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIGLEC8 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

SIGLEC8 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It preferentially binds to alpha2,3-linked sialic acid. and also binds to alpha2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. SIGLEC8 is expressed specifically on eosinophils. The protein contains 1 copy of a cytoplasmic motif that is referred to as the immunoreceptor tyrosine-based inhibitor motif (ITIM). This motif is involved in downmodulation of cellular responses. The phosphorylated ITIM motif binds to the SH2 domain of PTPN6/SHP-1. The SIGLEC8 gene belongs to the immunoglobulin superfamily.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC8
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1A5B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SIGLEC8
Gene Accession No. Q9NYZ4
Gene Alias CDw329; MGC59785; SAF2; SAF-2; sialic acid binding Ig like lectin 8; sialic acid binding Ig-like lectin 8; sialic acid-binding Ig-like lectin 8; Sialoadhesin family member 2; Siglec; SIGLEC8; Siglec-8; SIGLEC8L
Gene Symbols SIGLEC8
Host Species Mouse
Immunogen Purified recombinant fragment of human SIGLEC8 (AA: extra 17-216) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27181, 292846
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.