Learn More
Invitrogen™ SIGLEC8 Polyclonal Antibody

Description
Immunogen sequence: MEGDRQYGDG YLLQVQELVT VQEGLCVHVP CSFSYPQDGW TDSDPVHGYW FRAGDRPYQD APVATNNPDR EVQAETQGRF QLLGDIWSND CSLSIRDARK RDKGSYFFRL ERGSMKWSYK SQLNYKTKQL SVFVTALTHR PDILILGTLE SGHSRNLTCS VPWACKQGTP PMISWIGASV SSPGPTTARS SVLTLTPKPQ DHGTSLTCQV TLPGTGVTTT STVRLDVSYP PWNLTMTVFQ GDATASTALG NGSSLSVLEG QSLRLVCAVN SNPPARLSWT RGSLTLCPSR SSNPGLLELP RVHVRDEGEF TCRAQNAQGS QHISLSLSLQ NEGTGTSRPV SQVTLAA; Positive Samples: HL-60, 293T, Mouse liver, Rat blood; Cellular Location: Membrane, Single-pass type I membrane protein.
Specifications
Specifications
| Antigen | SIGLEC8 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SIGLEC8 |
| Gene Accession No. | Q9NYZ4 |
| Gene Alias | CDw329; MGC59785; SAF2; SAF-2; sialic acid binding Ig like lectin 8; sialic acid binding Ig-like lectin 8; sialic acid-binding Ig-like lectin 8; Sialoadhesin family member 2; Siglec; SIGLEC8; Siglec-8; SIGLEC8L |
| Gene Symbols | SIGLEC8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.