Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIGLEC8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110774
Description
Immunogen sequence: RSCRKKSARP AAGVGDTGME DAKAIRGSAS QGPLTESWKD GNPLKKPPPA VAPSSGEEGE LHYATLSFHK VKPQDPQGQE ATDSEYSEIK IHKRETAETQ ACLRNHNPSS KEVRG.
SIGLEC8 is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It preferentially binds to alpha2,3-linked sialic acid. and also binds to alpha2,6-linked sialic acid. The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. SIGLEC8 is expressed specifically on eosinophils. The protein contains 1 copy of a cytoplasmic motif that is referred to as the immunoreceptor tyrosine-based inhibitor motif (ITIM). This motif is involved in downmodulation of cellular responses. The phosphorylated ITIM motif binds to the SH2 domain of PTPN6/SHP-1. The SIGLEC8 gene belongs to the immunoglobulin superfamily.
Specifications
SIGLEC8 | |
Polyclonal | |
Unconjugated | |
SIGLEC8 | |
CDw329; MGC59785; SAF2; SAF-2; sialic acid binding Ig like lectin 8; sialic acid binding Ig-like lectin 8; sialic acid-binding Ig-like lectin 8; Sialoadhesin family member 2; Siglec; SIGLEC8; Siglec-8; SIGLEC8L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27181 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NYZ4 | |
SIGLEC8 | |
Recombinant Protein corresonding to Human SIGLEC8. Recombinant protein control fragment (Product #RP-90052). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction