Learn More
Invitrogen™ SIGLEC9 Monoclonal Antibody (OTI8F7)

Description
SIGLEC9 Monoclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | SIGLEC9 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI8F7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SIGLEC9 |
| Gene Accession No. | Q9Y336 |
| Gene Alias | Cd170; CD329; CDw329; FOAP-9; LOC100062855; LOC719831; MIS; mSiglec-E; Myeloid inhibitory siglec; OBBP-LIKE; Protein FOAP-9; sialic acid binding Ig like lectin 9; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin 9; sialic acid binding Ig-like lectin E; sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 5; sialic acid-binding Ig-like lectin 9; Sialic acid-binding Ig-like lectin E; sialic acid-binding Ig-like lectin-like 1; sialic acid-binding immunoglobulin-like lectin E; Siglec; Siglec12; siglec-12; Siglec5; Siglec-5; SIGLEC9; siglec-9; Siglece; Siglec-E; Siglecl1; siglec-L1; SIGLEC-like 1; UNQ668/PRO1302 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.