Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGLEC9 Mouse anti-Human, Clone: OTI1D9, liquid, TrueMAB™
SDP

Catalog No. 501671512
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1512 Supplier OriGene Supplier No. TA500379
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Siglec9 (Sialic acid-binding Ig-like lectin 9) is single-pass type I membrane protein and belongs to immunoglobulin super family and contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It is reported that Siglec9 recognizes sialic acid in alpha2, 3-or alpha2, 6-glycosidic linkage to galactose. Using a specific antibody, Siglec9 was found to be expressed at high or intermediate levels by monocytes, neutrophils, and a minor population of CD16+, CD56- cells. Siglec9 also acts as an inhibitory receptor for MUC16 that attenuates T- andNK cell function. Recent reports suggest that Siglec9 expressed on DCs is involved in immuno regulation through ligation with mucins in an epithelial cancer patient.

Specifications

Antigen SIGLEC9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D9
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SIGLEC9
Gene Accession No. Q9Y336
Gene Alias CD329, CDw329, FOAP-9, OBBP-LIKE, siglec-9
Gene Symbols SIGLEC9
Host Species Mouse
Immunogen Full length human recombit protein of human SIGLEC9 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27180
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.