Learn More
SIGLEC9 Mouse anti-Human, Clone: OTI6D8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500382

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Siglec9 (Sialic acid-binding Ig-like lectin 9) is single-pass type I membrane protein and belongs to immunoglobulin super family and contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It is reported that Siglec9 recognizes sialic acid in alpha2, 3-or alpha2, 6-glycosidic linkage to galactose. Using a specific antibody, Siglec9 was found to be expressed at high or intermediate levels by monocytes, neutrophils, and a minor population of CD16+, CD56- cells. Siglec9 also acts as an inhibitory receptor for MUC16 that attenuates T- andNK cell function. Recent reports suggest that Siglec9 expressed on DCs is involved in immuno regulation through ligation with mucins in an epithelial cancer patient.Specifications
SIGLEC9 | |
Monoclonal | |
Unconjugated | |
SIGLEC9 | |
CD329, CDw329, FOAP-9, OBBP-LIKE, siglec-9 | |
Mouse | |
Affinity Chromatography | |
RUO | |
27180 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI6D8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y336 | |
SIGLEC9 | |
Full length human recombit protein of human SIGLEC9 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.