Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIGLEC9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582404
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582404 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582404 Supplier Invitrogen™ Supplier No. PA582404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QNLTMTVFQG DGTVSTVLGN GSSLSLPEGQ SLRLVCAVDA VDSNPPARLS LSWRGLTLCP SQPSNPGVLE LPWVHLRDEA EFTCRAQNPL GSQQVYLNVS LQSKATSGVT Q Highest antigen sequence indentity to the following orthologs: Rat - 55%, Mouse - 55%.

SIGLEC9 (Sialic acid-binding Ig-like lectin 9) is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It preferentially binds to alpha2, 3-or alpha2, 6-linked sialic acid. The sialic acid recognition site may be mased by cis interactions with sialic acids on the same cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIGLEC9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SIGLEC9
Gene Accession No. Q9Y336
Gene Alias Cd170; CD329; CDw329; FOAP-9; LOC100062855; LOC719831; MIS; mSiglec-E; Myeloid inhibitory siglec; OBBP-LIKE; Protein FOAP-9; sialic acid binding Ig like lectin 9; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin 9; sialic acid binding Ig-like lectin E; sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 5; sialic acid-binding Ig-like lectin 9; Sialic acid-binding Ig-like lectin E; sialic acid-binding Ig-like lectin-like 1; sialic acid-binding immunoglobulin-like lectin E; Siglec; Siglec12; siglec-12; Siglec5; Siglec-5; SIGLEC9; siglec-9; Siglece; Siglec-E; Siglecl1; siglec-L1; SIGLEC-like 1; UNQ668/PRO1302
Gene Symbols SIGLEC9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SIGLEC9. Recombinant protein control fragment (Product #RP-90194).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27180
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.