Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIGLEC9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110371
Description
Immunogen sequence: SLTCQVTFPG ASVTTNKTVH LNVSYPPQNL TMTVFQGDGT VSTVLGNGSS LSLPEGQSLR LVCAVDAVDS NPPARLSLSW RGLTLCPSQP SNPGVLELPW VHLRDAAEFT CRAQNPLGSQ QVYLNVSLQS K.
SIGLEC9 (Sialic acid-binding Ig-like lectin 9) is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It preferentially binds to alpha2, 3-or alpha2, 6-linked sialic acid. The sialic acid recognition site may be mased by cis interactions with sialic acids on the same cell surface.
Specifications
| SIGLEC9 | |
| Polyclonal | |
| Unconjugated | |
| SIGLEC9 | |
| Cd170; CD329; CDw329; FOAP-9; LOC100062855; LOC719831; MIS; mSiglec-E; Myeloid inhibitory siglec; OBBP-LIKE; Protein FOAP-9; sialic acid binding Ig like lectin 9; sialic acid binding Ig-like lectin 5; sialic acid binding Ig-like lectin 9; sialic acid binding Ig-like lectin E; sialic acid-binding Ig-like lectin 12; Sialic acid-binding Ig-like lectin 5; sialic acid-binding Ig-like lectin 9; Sialic acid-binding Ig-like lectin E; sialic acid-binding Ig-like lectin-like 1; sialic acid-binding immunoglobulin-like lectin E; Siglec; Siglec12; siglec-12; Siglec5; Siglec-5; SIGLEC9; siglec-9; Siglece; Siglec-E; Siglecl1; siglec-L1; SIGLEC-like 1; UNQ668/PRO1302 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27180 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.04 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9Y336 | |
| SIGLEC9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 210-340 of human SIGLEC9 (NP_001185487.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction