Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIGLEC9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$518.60

Specifications

Antigen SIGLEC9
Concentration 0.36 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-8693
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
133771AP150UL
150 μL
Each of 1 for $518.60
Only null left
Add to Cart
 
Description

Description

Siglec9 (Sialic acid-binding Ig-like lectin 9) is single-pass type I membrane protein and belongs to immunoglobulin super family and contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It is reported that Siglec9 recognizes sialic acid in alpha2, 3-or alpha2, 6-glycosidic linkage to galactose. Using a specific antibody, Siglec9 was found to be expressed at high or intermediate levels by monocytes, neutrophils, and a minor population of CD16+, CD56- cells. Siglec9 also acts as an inhibitory receptor for MUC16 that attenuates T- andNK cell function. Recent reports suggest that Siglec9 expressed on DCs is involved in immuno regulation through ligation with mucins in an epithelial cancer patient.
Specifications

Specifications

SIGLEC9
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse
Q9Y336
27180, 83382
SIGLEC9 Fusion Protein Ag4201
Primary
-20°C
SIGLEC9
0.36 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CD329, CDw329, OBBP LIKE, Protein FOAP 9, Siglec 9, SIGLEC9
SIGLEC9, Siglece
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.