Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIGLECL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546369
Description
Peptide sequence: RKKQAKKAAA IRAKKSSKVR ASQELEMSLK PEEPGKPVVA TFSESRILEK Sequence homology: Human: 100%; Yeast: 83%.
The function of this protein remains unknown.
Specifications
SIGLECL1 | |
Polyclonal | |
Unconjugated | |
SIGLECL1 | |
C19orf75; sialic acid binding Ig-like lectin, pseudogene 7; Siglec; SIGLEC family like 1; SIGLEC family-like protein 1; SIGLEC23P; SIGLECL1; SIGLECP7 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
284369 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N7X8 | |
SIGLECL1 | |
Synthetic peptide directed towards the C-terminal of human SIGLECL1 (aa 142-191). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction