Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sigma-1 Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530372 Supplier Invitrogen™ Supplier No. PA530372
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T. Predicted reactivity: Mouse (94%), Rat (96%), Rhesus Monkey (100%), Bovine (97%), Guinea pig (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a receptor protein that interacts with a variety of psychotomimetic drugs, including cocaine and amphetamines. The receptor is believed to play an important role in the cellular functions of various tissues associated with the endocrine, immune, and nervous systems. As indicated by its previous name, opioid receptor sigma 1 (OPRS1), the product of this gene was erroneously thought to function as an opioid receptor; it is now thought to be a non-opioid receptor. Alternative splicing of this gene results in transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sigma-1 Receptor
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SIGMAR1
Gene Accession No. O55242, Q7ZWG9, Q99720
Gene Alias AAG8; Aging-associated gene 8 protein; AL024364; ALS16; DSMA2; hSigmaR1; mSigmaR1; opioid receptor, sigma 1; Oprs1; RP23 167I12.6; SIG 1R; SIG-1R; sigma 1 receptor variant SR-1D; sigma 1-type opioid receptor; Sigma non-opioid intracellular receptor 1; sigma1 receptor; Sigma-1 Receptor; sigma1R; sigma1-receptor; Sigmar1; SR BP; SR31747 binding protein 1; SR31747-binding protein; SRBP; SR-BP; SR-BP1; zgc:56378
Gene Symbols SIGMAR1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SIGMAR1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10280, 18391, 393952
Target Species Human, Mouse, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.