Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sigma 54 Monoclonal Antibody (33.7)
GREENER_CHOICE

Catalog No. p-7178171
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-954 100 μL
01-674-955 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-674-954 Supplier Invitrogen™ Supplier No. MA15876
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody binds specifically in ELISA to monomeric tetrameric Sigma 54 but it does not bind to RAT protein or AgrA protein.

Sigma factors are initiation factors that promote the attachment of RNA polymerase to specific initiation sites and are then released. This sigma factor is responsible for the expression of enzymes involved in arginine catabolism. The open complex (sigma54 and core RNA polymerase) serves as the receptor for the receipt of the melting signal from the remotely bound activator protein glnG(ntrC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sigma 54
Applications ELISA, Western Blot
Classification Monoclonal
Clone 33.7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Host Species Mouse
Immunogen Bacterially expressed Sigma 54 from Balb/c Sp2/0 cells, purified over a heparin column.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.