Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Eosin Y Solution, Alcoholic with Phloxine

Catalog No. p-2386937
Encompass
Click to view available options
Quantity:
500 mL
1 L

General-purpose cytoplasmic counterstain

  • Certified Eosin Y, 0.1% (w/v) and certified phloxine B, 0.1% (w/v) in acidified ethanol
TRUSTED_SUSTAINABILITY

Specifications

Color Bright pink to pinkish-orange
Type Alcoholic Eosin Y Stain
Format Solution
For Use With (Application) Used with hematoxylin and eosin staining
Product Type Eosin Y solution, alcoholic, with phloxine
Quantity 500 mL
Storage Requirements 18–26°C
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.