Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Periodic Acid Schiff Kit (PAS) Kit

Catalog No. sd395b
Encompass
Click to view available options
Quantity:
1 Kit

Includes

  • Hematoxylin Solution, Gill No. 3 (kit only) 100mL
  • Periodic Acid Solution (395-1) 2 x 100mL
  • Schiff's Reagent (kit only) 4 x 50mL

For demonstration of lymphocytes and mucopolysaccharides in tissue sections

 

  • PAS staining procedure may also be used for the demonstration of fungal organisms in tissue sections
  • Reaction can be performed on blood or bone marrow films, tissue touch preparations, or tissue sections
  • For in vitro diagnostic use
TRUSTED_SUSTAINABILITY

Specifications

Type Periodic Acid Schiff
For Use With (Application) Demonstration of lymphocytes and mucopolysaccharides
Includes
  • Hematoxylin Solution, Gill No. 3 (kit only) 100mL
  • Periodic Acid Solution (395-1) 2 x 100mL
  • Schiff's Reagent (kit only) 4 x 50mL
Product Type Periodic Acid-Schiff (PAS) Kit
Quantity 1 Kit
Storage Requirements 2-8°C
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.