Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Signal Peptide Peptidase Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232276F
Description
Signal Peptide Peptidase Polyclonal antibody specifically detects Signal Peptide Peptidase in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Signal Peptide Peptidase | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| dJ324O17.1, EC 3.4.23.-, H13hIMP1, histocompatibility (minor) 13, IMP-1, IMP1MSTP086, Intramembrane protease 1, minor histocompatibility antigen 13, minor histocompatibility antigen H13, Presenilin-like protein 3, PSENL3, PSL3IMPAS, Signal peptide peptidase, signal peptide peptidase beta, SPPIMPAS-1 | |
| The immunogen this antibody was made to, maps to a region between residue 327 to 377 of human Signal Peptide Peptidase Like 1 using the numbering given in entry NP_110416.1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 81502 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction