Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Signal peptide peptidase-like 3, Mouse anti-Human, Clone: 7F9, Millipore Sigma™

Catalog No. MABS191025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS191025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS191025 Supplier MilliporeSigma Supplier No. MABS191025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Signal peptide peptidase-like 3, clone 7F9, Cat. No. MABS1910, is a mouse monoclonal antibody that detects Signal peptide peptidase-like 3 and has been tested for use in Immunoprecipitation and Western Blotting.

Signal peptide peptidase-like 3 (UniProt: Q8TCT6; also known as SPP-like 3, Intramembrane protease 2, IMP-2, Presenilin homologous protein 1, PSH1, Presenilin-like protein 4, SPPL3) is encoded by the SPPL3 (also known as IMP2) gene (Gene ID: 121665) in human. SPPL3 is a multi-pass transmembrane protein that is shown to be widely expressed and is highly conserved in multicellular eukaryotes. Three isoforms of SPPL3 have been described that are produced by alternative splicing. It is a member of the intramembrane-cleaving GxGD proteases. It acts as an intramembrane protease that cleaves type II membrane protein substrates in or close to their luminal transmembrane domain boundaries. It can also act as a sheddase by mediating the proteolytic release and secretion of active site-containing ectodomains of glycan-modifying glycosidases and glycosyltransferases. It is also shown to catalyze the intramembrane cleavage of the envelope glycoprotein gp130 and/or the leader peptide gp18LP of the simian foamy virus independent of prior ectodomain shedding by furin or furin-like proprotein convertase-mediated cleavage proteolysis. The first transmembrane domain of SPPL3 may act as a type I signal anchor. SPPL3 also contains a PAL motif (aa 342-344) that is required for normal active site conformation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Signal peptide peptidase-like 3
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 7F9
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected Signal peptide peptidase-like 3 in Western Blotting applications (Voss, M., et. al. (2012). J Biol Chem. 287(52):43401-9; Voss, M., et. al. (2014). EMBO J. 33(24):2890-905; Fleck, D., et. al. (2016). J Biol Chem. 291(1):318-33).Immunoprecipitation Analysis: A representative lot detected Signal peptide peptidase-like 3 in Immunoprecipitation applications (Voss, M., et. al. (2012). J Biol Chem. 287(52):43401-9).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Signal peptide peptidase-like 3;SPP-like 3;Intramembrane protease 2;IMP-2;Presenilin homologous protein 1;PSH1;Presenilin-like protein 4
Gene Symbols SPPL3;IMP2
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human Signal peptide peptidase-like 3.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 7F9 detects Signal peptide peptidase-like 3 in human cells. it targets an epitope within 15 amino acids from the C-terminal half. Immunogen sequence is conserved in all three isoforms.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.