Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Signal sequence receptor delta Antibody (26G5), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242656AF647
Description
Signal sequence receptor delta Monoclonal specifically detects Signal sequence receptor delta in Human samples. It is validated for Western Blot, ELISA.Specifications
Signal sequence receptor delta | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SSR4 | |
Recombinant human SSR4 (24-144aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
26G5 | |
Western Blot, ELISA | |
signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta | |
Mouse | |
Protein A purified | |
RUO | |
6748 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction