Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Signal sequence receptor delta Antibody (26G5), Alexa Fluor™ 750, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242656AF750

Catalog No. NP24266A70


Only null left
Add to Cart

Description

Description

Signal sequence receptor delta Monoclonal specifically detects Signal sequence receptor delta in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Signal sequence receptor delta
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
SSR4
Recombinant human SSR4 (24-144aa) purified from E. coli
Primary
Human
IgG1 κ
Western Blot, ELISA
26G5
Western Blot, ELISA
signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta
Mouse
100 μL
6748
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.