Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Signal sequence receptor delta Antibody (26G5), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242656MFV450
Description
Signal sequence receptor delta Monoclonal antibody specifically detects Signal sequence receptor delta in Human samples. It is validated for Western Blot, ELISASpecifications
| Signal sequence receptor delta | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 26G5 | |
| Western Blot, ELISA | |
| signal sequence receptor, delta (translocon-associated protein delta), SSR-delta, translocon-associated protein delta, translocon-associated protein subunit delta, TRAP-delta, TRAPDSignal sequence receptor subunit delta | |
| Recombinant human Signal sequence receptor delta (24-144aa) purified from E. coli | |
| 0.1 mL | |
| Signal Transduction | |
| 6748 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction