Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIGNR1/CD209b Antibody (123), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290608APC
Description
SIGNR1/CD209b Monoclonal antibody specifically detects SIGNR1/CD209b in Mouse samples. It is validated for ELISA, ELISASpecifications
SIGNR1/CD209b | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
170786 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
123 | |
ELISA, Sandwich ELISA Capture | |
CD209 antigen like protein B, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
Rabbit immunized with purified, recombinant Mouse SIGNR1/CD209b (Accession#: NP_081248.2, Gln74-Gly325) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction