Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIGNR1/CD209b Antibody (143), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290609R
Description
SIGNR1/CD209b Monoclonal specifically detects SIGNR1/CD209b in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
SIGNR1/CD209b | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
170786 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunofluorescence | |
143 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
CD209 antigen like protein B, DC SIGN related protein 1, DC SIGNR1, OtB7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIGNR1/CD209b (NP_081248.2; Gln74-Gly325). | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction