Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIGNR4/CD209e Antibody (RM0327-9B29), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212431AF350
Description
SIGNR4/CD209e Monoclonal specifically detects SIGNR4/CD209e in Mouse samples. It is validated for Western Blot.Specifications
SIGNR4/CD209e | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
Mouse recombinant SIGNR4/CD209e | |
0.1 mL | |
Mouse | |
IgG2 |
Western Blot | |
RM0327-9B29 | |
Western Blot | |
Rat | |
Protein A or G purified | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction