Learn More
Promega SIK Kinase Enzyme System, 10microg
Catalog No. PRV4156 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4156 10 μg
1 options
Catalog No. PRV4156 Supplier Promega Supplier No. V4156
Recombinant human SIK (1-303) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. SIK is a protein kinase that is involved in regulating AMPK-related kinases.
  • Easily Screen and Profile SIK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C