Learn More
Promega SIK Kinase Enzyme System, 1mg
Catalog No. PRV6561 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6561 1 mg
1 options
Catalog No. PRV6561 Supplier Promega Supplier No. V6561
Add to Cart
Add to Cart
Request a Quote
Recombinant human SIK (1-303) was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. SIK is a protein kinase that is involved in regulating AMPK-related kinases.
  • Easily Screen and Profile SIK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C