Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIK1 Polyclonal Antibody, PE, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6252RPE
This item is not returnable.
View return policy
Description
SIK1 Polyclonal antibody specifically detects SIK1 in Human, Mouse samples. It is validated for Flow CytometrySpecifications
| SIK1 | |
| Polyclonal | |
| PE | |
| SIK1 | |
| EIEE30; HRT-20; Kid2; MSK; Myocardial SNF1-like kinase; Probable serine/threonine protein kinase SNF1LK (HRT-20) (Myocardial SNF1-like kinase); protein kinase KID2; salt inducible kinase 1; salt-inducible kinase 1; salt-inducible protein kinase 1; serine/threonine protein kinase; serine/threonine-protein kinase SIK1; Serine/threonine-protein kinase SNF1-like kinase 1; serine/threonine-protein kinase SNF1LK; SIK; Sik1; SIK-1; SNF1-like kinase; SNF1LK | |
| KLH conjugated synthetic peptide derived from human Snf1lk | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
| P57059, Q60670 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 150094, 17691 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction