Learn More
Invitrogen™ SIK2 Polyclonal Antibody

Description
Immunogen sequence: HFAAIYFLLV ERLKSHRSSF PVEQRLDGRQ RRPSTIAEQT VAKAQTVGLP VTMHSPNMRL LRSALLPQAS NVEAFSFPAS GCQAEAAFME EECVDTPKVN GCLLDPVPPV LVRKGCQSLP SNMMETSIDE GLE Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 94%.
Specifications
Specifications
| Antigen | SIK2 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SIK2 |
| Gene Accession No. | Q9H0K1 |
| Gene Alias | G630080D20Rik; KIAA0781; LOH11CR1I; QIK; Qin-induced kinase; RGD1559698; salt induceable kinase 2; salt inducible kinase 2; salt-inducible kinase 2; salt-inducible protein kinase 2; salt-inducible serine/threonine kinase 2; serine/threonine-protein kinase SIK2; Serine/threonine-protein kinase SNF1-like kinase 2; Sik2; SIK-2; SNF1-like kinase 2; SNF1LK2 |
| Gene Symbols | SIK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.