Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIK3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$382.00 - $648.50
Specifications
Antigen | SIK3 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
SIK3 Polyclonal specifically detects SIK3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SIK3 | |
Polyclonal | |
Rabbit | |
Protein Kinase | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
23387 | |
This antibody was developed against a recombinant protein corresponding to amino acids: QQEYQELFRHMNQGDAGSLAPSLGGQSMTERQALSYQNADSYHHHTSPQHLLQIRAQECVSQASSPTPPHGYAHQPALMHSESMEEDCSCEGAKDGFQD | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Mouse | |
EC 2.7.11, EC 2.7.11.1, KIAA0999L19, QSKFLJ12240, Salt-inducible kinase 3, Serine/threonine-protein kinase QSK, serine/threonine-protein kinase SIK3, SIK family kinase 3, SIK-3 | |
SIK3 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title