Learn More
Invitrogen™ SIK3 Polyclonal Antibody

Description
Immunogen sequence: QQEYQELFRH MNQGDAGSLA PSLGGQSMTE RQALSYQNAD SYHHHTSPQH LLQIRAQECV SQASSPTPPH GYAHQPALMH SESMEEDCSC EGAKDGFQD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 80%.
Specifications
Specifications
| Antigen | SIK3 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SIK3 |
| Gene Accession No. | Q9Y2K2 |
| Gene Alias | 5730525O22Rik; 9030204A07Rik; AI447252; AI846254; AI849445; Kiaa0999; L19; mKIAA0999; QSK; Salt-inducible kinase 3; serine/threonine-protein kinase QSK; Serine/threonine-protein kinase SIK3; SIK family kinase 3; SIK3; SIK-3 |
| Gene Symbols | SIK3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.