Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIK3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567661
Description
Predicted to react with Mouse and Rat samples.
SIK3 (QSK) is a serine/threonine-protein kinase, belongs to QIK subfamily. The phosphorylation of SIK3 by LKB1 through the 14-3-3 binding enhances its catalytic activity and leads its localization to punctate structures within the cytoplasm. Overexpression of SIK3 promotes G1/S cell cycle progression with ovarian cancer. There are two sites (H331L and A1103V) were mutated at significant frequency in breast cancer. SIK3 is a novel tumor-associated antigen (TAA). SIK3 is a positive regulator of mTOR signaling that functions by triggering the degradation of DEPTOR, an mTOR inhibitor. Involved in the dynamic regulation of mTOR signaling in chondrocyte differentiation during skeletogenesis. Negatively regulates cAMP signaling pathway possibly by acting on CRTC2/TORC2 and CRTC3/TORC3.
Specifications
SIK3 | |
Polyclonal | |
Unconjugated | |
SIK3 | |
5730525O22Rik; 9030204A07Rik; AI447252; AI846254; AI849445; Kiaa0999; L19; mKIAA0999; QSK; Salt-inducible kinase 3; serine/threonine-protein kinase QSK; Serine/threonine-protein kinase SIK3; SIK family kinase 3; SIK3; SIK-3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23387, 684112, 70661 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 30% glycerol and 0.01% sodium azide; pH 7.3 | |
Q6P4S6, Q9Y2K2 | |
SIK3 | |
Synthesized peptide derived from human SIK3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction