Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIKE Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555247
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555247 Supplier Invitrogen™ Supplier No. PA555247
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEPVLKAHQS HSAEIESQID RICEMGEVMR KAVQVDDDQF CKIQEKLAQL ELENKELREL LSISSESLQA RKENSMD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.

Physiological suppressor of IKK-epsilon and TBK1 that plays an inhibitory role in virus- and TLR3-triggered IRF3. Inhibits TLR3-mediated activation of interferon-stimulated response elements (ISRE) and the IFN-beta promoter. May act by disrupting the interactions of IKBKE or TBK1 with TICAM1/TRIF, IRF3 and DDX58/RIG-I. Does not inhibit NF-kappa-B activation pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIKE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SIKE1
Gene Accession No. Q9BRV8
Gene Alias 2810005O12Rik; 5730470L24Rik; AI450236; AI839862; RGD1311316; SIKE; SIKE1; Sikeb; suppressor of IKBKE 1; suppressor of IKK epsilon; suppressor of IKK-epsilon
Gene Symbols SIKE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SIKE. Recombinant protein control fragment (Product #RP-93990).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80143
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.