Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115974
Description
Antibody detects endogenous levels of total STIL.
Plays an essential role in early embryonic development.
Specifications
| SIL | |
| Polyclonal | |
| Unconjugated | |
| STIL | |
| cassiopeia; chunp6907; csp; MCPH7; nbb; scl/tal1 interrupting locus; SCL-interrupting locus protein; SCL-interrupting locus protein homolog; SIL; sill; Stil; STIL centriolar assembly protein; stil protein; TAL1 (SCL) interrupting locus like; Tal1 interrupting locus; TAL1/SCL interrupting locus-like protein; TAL-1-interrupting locus protein; zgc:110502 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20460, 313506, 6491 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q15468, Q60988 | |
| STIL | |
| A synthesized peptide derived from human STIL(Accession Q15468), corresponding to amino acid residues E1236-L1286. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction