Learn More
SIL Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302442AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Plays an essential role in early embryonic development.Specifications
SIL | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q15468, Q60988 | |
Stil | |
Between 1237 and 1287 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
STIL | |
SCL-interrupting locus protein homolog, stil, sill | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20460, 6491 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.