Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIL1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001304
Description
SIL1 Polyclonal specifically detects SIL1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
SIL1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
BAPMarinesco-Sjogren syndrome, BiP-associated protein, MSS, nucleotide exchange factor SIL1, SIL1 homolog, endoplasmic reticulum chaperone (S. cerevisiae), ULG5 | |
Goat | |
Affinity Purified | |
RUO | |
64374 | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 2 ug/ml, Peptide ELISA Detection limit 1:8000 | |
NP_071909. | |
SIL1 | |
Peptide with sequence C-ELLGSVNSLLKELR corresponding to C-Terminus according to NP_071909.1, NP_001032722.1. | |
0.1 mg | |
Primary | |
Variants (NP_071909.1; NP_001032722.1) encode the same isoform. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction