Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIL1 Monoclonal Antibody (OTI3B11), TrueMAB™, OriGene
SDP

Catalog No. 502542608
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2608 30 μL
50-167-2153 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2608 Supplier OriGene Supplier No. TA501010S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIL1 Monoclonal antibody specifically detects SIL1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen SIL1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B11
Concentration 0.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene TUBA8
Gene Accession No. Q9NY65
Gene Alias 1810057E01Rik; AI042831; BAP; BiP-associated protein; endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae); homolog of yeast; MSS; Nucleotide exchange factor SIL1; SIL1; SIL1 homolog, endoplasmic reticulum chaperone; SIL1 nucleotide exchange factor; SIL1 protein; SIL1-like protein endoplasmic reticulum chaperone; ULG5; UNQ545/PRO836; wz
Host Species Mouse
Immunogen Full length human recombinant protein of human TUBA8 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51807
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.