Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIL1 Monoclonal Antibody (OTI3E3)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA525059

Catalog No. PIMA525059


Only null left
Add to Cart

Description

Description

SIL1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome. Alternate transcriptional splice variants have been characterized.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIL1
Monoclonal
0.56 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H173
SIL1
Full length human recombinant protein of SIL1 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI3E3
Unconjugated
SIL1
1810057E01Rik; AI042831; BAP; BiP-associated protein; endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae); homolog of yeast; MSS; Nucleotide exchange factor SIL1; SIL1; SIL1 homolog, endoplasmic reticulum chaperone; SIL1 nucleotide exchange factor; SIL1 protein; SIL1-like protein endoplasmic reticulum chaperone; ULG5; UNQ545/PRO836; wz
Mouse
Affinity Chromatography
RUO
64374
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.