Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530799
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530799 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530799 Supplier Invitrogen™ Supplier No. PA530799
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, rat liver. Predicted reactivity: Mouse (92%), Rat (93%), Chimpanzee (98%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome. Alternate transcriptional splice variants have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIL1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SIL1
Gene Accession No. Q6P6S4, Q9H173
Gene Alias 1810057E01Rik; AI042831; BAP; BiP-associated protein; endoplasmic reticulum chaperone SIL1 homolog (S. cerevisiae); homolog of yeast; MSS; Nucleotide exchange factor SIL1; SIL1; SIL1 homolog, endoplasmic reticulum chaperone; SIL1 nucleotide exchange factor; SIL1 protein; SIL1-like protein endoplasmic reticulum chaperone; ULG5; UNQ545/PRO836; wz
Gene Symbols SIL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SIL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291673, 64374
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.