Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ SILAC DMEM Flex Media, no glucose, no phenol red

Catalog No. A2493901
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2493901 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2493901 Supplier Gibco™ Supplier No. A2493901
Add to Cart
Add to Cart

SILAC DMEM Flex Media, no glucose, no phenol red

SILAC DMEM Flex is Dulbecco’s Modified Eagle Medium (DMEM) without L-glutamine, L-lysine, and L-arginine for use during SILAC protein labeling with stable isotopic labeled lysine and/or arginine. DMEM is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM include primary fibroblasts, neurons, glial cells, HUVECs, and smooth muscle cells, as well as cell lines such as HeLa, 293, Cos-7, and PC-12. We offer a variety of Gibco™ DMEM modifications for a range of applications. Find the right modification using the media selector tool. DMEM was originally formulated with low glucose (1 g/L) and sodium pyruvate, but is often used with higher glucose levels, with or without sodium pyruvate. SILAC DMEM Flex has been further modified to not contain lysine, arginine, and glutamine. DMEM contains no proteins, lipids, or growth factors. Therefore, DMEM requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). SILAC protein labeling experiments should be conducted using Gibco™ Dialyzed FBS. DMEM uses a sodium bicarbonate buffer system (3.7 g/L), so requires a 5-10% CO2 environment to maintain physiological pH.

cGMP Manufacturing and Quality System
SILAC DMEM Flex is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Mammalian Cells
Form Liquid
Product Type Dulbecco's Modified Eagle Medium
Serum Level Standard Serum Supplementation
Without Additives No Glucose, No Glutamine, No HEPES, No Phenol Red, No Sodium Pyruvate, No Arginine, No Lysine
Product Line Gibco
Quantity 500 mL
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Shelf life: 12 months from date of manufacture.
Culture Type Mammalian Cell Culture
Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

For Research Use or Further Manufacturing. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.