Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sildenafil Citrate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18606195
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle

S3092, 171599-83-0, C22H30N6O4S·C6H8O7

Spectrum™ Chemical Sildenafil Citrate, USP

Chemical Identifiers

CAS 171599-83-0
Molecular Formula C28H38N6O11S
Molecular Weight (g/mol) 666.70
InChI Key DEIYFTQMQPDXOT-UHFFFAOYSA-N
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid; 5-{2-ethoxy-5-[(4-methylpiperazin-1-yl)sulfonyl]phenyl}-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC(=CC=C1OCC)S(=O)(=O)N1CCN(C)CC1

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.