Learn More
SiliCycle™ SiliaBond™ Functionalized Silica Gel Carbonate (CO3), 40 to 63 μm, 60 Å
Our Carbonate Silica Gel is a powerful scavenger for the removal of acids, acidic phenols and boronic acids.
Supplier: SiliCycle™ R66030B500G
Description
SiliaBond™ Carbonate acts as an Organic Scavenger for Acids (Ionic Bonding and Catch and Release for Acids, Acidic Phenols and Boronic Acids). You only need to filter out the scavenger once it has trapped the residual metal or organic impurity. The gel is great for a variety of formats for all purifications scales, from laboratory to process to plant scales. It is a successful technology for pharmaceuticals, organic chemistry labs, agrochemicals, mining, fine chemicals, water and waste treatment, and offers great compatibility with a myriad of experimental conditions, solvents, pH and temperatures.
- Strong chemical, physical, thermal and mechanical stability
- Made in accordance with sustainable process chemistry and green metrics
- No hazards; no safety precautions needed
Specifications
Bulk Functionalized Irregular Silica Gel | |
500 g | |
SiliaBond™ |
Irregular | |
HDPE Bottle | |
0.608 g/mL |
Safety and Handling
Recommended Storage : Keep Dry
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.