Learn More
SiliCycle™ SiliaSphere™ C18, 10 μm, 60 Å
Catalog No. p-7238447
SiliaSphere™ C18, 10 μm, 60 Å is a great start for method development as it presents the maximum retention of non-polar compounds.
  • Usually used for peptides, pesticides, PCBs, PAHs, drugs, etc.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.