Learn More
Silica gel 60, 0.040-0.063mm (230-400 mesh), 99.5% (metals basis)
Description
Column chromatographySilica gel is used in column chromatography without applied pressure (gravity pulled). It is used as an adsorbent in various applications and in removing impurities. It is used as stationary base in column chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder, -230+400 Mesh,S.A. 500-600m |
Mesh Size | 230-400 Mesh |
Odor | Odorless |
Assay Percent Range | 99.5% (metals basis) |
Color | White |
Quantity | 5 kg |
Solubility Information | Soluble in hydrofluoric acid. Insoluble in water. |
IUPAC Name | dioxosilane |
Chemical Name or Material | Silica gel 60 |
Safety and Handling
Hazard Category | H373: May cause damage to organs through prolonged or repeated exposure. |
EINECSNumber | 231-545-4 |
RTECSNumber | VV7565000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.