Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, For Chromatography40-63 Micron (230-400 Mesh), 60A, Ultrapure, Spectrum™ Chemical
SDP

Catalog No. 18606201 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

SIL19, 7631-86-9

Spectrum™ Chemical Silica Gel, For Chromatography40-63 Micron (230-400 Mesh), 60A, Ultrapure is a porous and granular form of silicon dioxide which is synthetically made from sodium silicate. In chemistry, the silica gel is used as a stationary phase in chromatography. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Specifications

Particle Size 40 to 63 μm
Grade Ultrapure
Packaging Amber Glass Bottle
pH 6.5 to 7.5
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.