Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel, functionalized, Amino-3, ca.1.4mmol/g, part.size 40-63 micron, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
71888-97-6
C9H23NO5Si2
281.455
MFCD03427643
JIBSXWZIQTVOKJ-UHFFFAOYSA-N
172930
dioxosilane;3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC.O=[Si]=O
71888-97-6
C9H23NO5Si2
281.455
MFCD03427643
JIBSXWZIQTVOKJ-UHFFFAOYSA-N
172930
dioxosilane;3-triethoxysilylpropan-1-amine
CCO[Si](CCCN)(OCC)OCC.O=[Si]=O
For solid support synthesis
Loading 1.2mmol/g min. (molecular)
Color White
Quantity 25g
Nitrogen (N) 1.68% min.
Packaging Glass bottle
Particle Size Particle size: 40 to 63 Micron
Physical Form Powder
Chemical Name or Material Silica gel, functionalized, Amino-3