Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel, High-Purity Grade, MilliporeSigma™ Supelco™

Catalog No. p-7187460 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 kg

Catalog No. 11-101-2554

Supplier: MilliporeSigma Supelco 607651KG

Only null left
Add to Cart
Add to Cart
Silica gel is a rigid 3D network of colloidal silica. It is exclusively used as a support material for the active titanium(III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel is classified as below:a. Aquagel - pores are filled with waterb. Xerogel - by the process of evaporation, aqueous phase in the pores are removedc. Aerogel - solvent is removed by supercritical extraction
TRUSTED_SUSTAINABILITY

Specifications

CAS 112926-00-8
Physical Form Powder
MDL Number MFCD00011232
Melting Point >1600°C
Boiling Point 2230°C
Type Organic Standard
Quantity 1 kg
Grade High Purity
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.