Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica gel desiccant, indicating, -6+16 mesh granules

Catalog No. p-5547816
Click to view available options
Quantity:
250 g
1 kg
5 kg

CAS: 7631-86-9 | O2Si | 60.08 g/mol

Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Color Blue
Physical Form Granular
Density 2.17 to 2.66g/cm3
Material Silica Gel
Quantity 250 g
Mesh Size -6+16 Mesh
Odor Odorless
Solubility Information Soluble in alkalies. Insoluble in water.
Sensitivity Hygroscopic
IUPAC Name dioxosilane
Grade ACS
Chemical Name or Material Silica gel desiccant
Show More Show Less
What color are the Silica gel desiccant, indicating, -6+16 mesh granules when they are activated or saturated?

Silica gel desiccant, indicating, -6+16 mesh granules gradually change from a deep blue in the activated condition to a pale pink in the saturated condition.

Can I regenerate the Silica gel desiccant, indicating, -6+16 mesh granules?

Yes, Silica gel desiccant, indicating, -6+16 mesh granules can be regenerated by heating at 121-232 degrees C until it returns to a blue color.

Hazard Category H350i-H360F
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 231-545-4
RTECSNumber VV7565000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.