Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Gel Rubin Drying Bags, Honeywell Fluka™

Catalog No. 60047446 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 kg
Packaging:
Metal Can
Poly Bottle

With indicator function, 1/3 TE, bag with ∼10 g, crystalline

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silica gel Rubin drying bags
Color Red
Boiling Point 2230°C
Melting Point 1600°C
pH 9
CAS 112926-00-8
Density 2g/cm3
Material Silica Gel
Quantity 1 kg
UN Number NONH for all modes of transport
Packaging Poly Bottle
Solubility Information Slightly soluble in water
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.