Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Silica gel, TLC high purity grade, with gypsum binder
Click to view available options
Quantity:
50 g
250 g
Description
TLC plate preparation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | 12 Micron APS,S.A. 500-600m |
| pH | 6 to 7 |
| Material | Silica Gel |
| Quantity | 50 g |
| Particle Size | 500 to 600m2/g, 60Å |
| Grade | TLC High Purity |
| Surface Area | 500 to 600m2/g |
| Chemical Name or Material | Silica gel, With gypsum binder |
Safety and Handling
| missing translation for 'healthHazard1' | Warning |
| missing translation for 'healthHazard2' | GHS H Statement H373 May cause damage to organs through prolonged or repeated exposure. |
| missing translation for 'healthHazard3' | GHS P Statement P260-P314-P501a Do not breathe dust/fume/gas/mist/vapors/spray. Get medical advice/attention if you feel unwell. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 231-545-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction