Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silica Standard, Certified, 1000ppm ±10ppm (1mL = 1mg SiO2), LabChem™

Catalog No. p-5388698
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle

Chemical Identifiers

CAS 13517-24-3, 7732-18-5
Molecular Formula H18Na2O12Si
Molecular Weight (g/mol) 284.197
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Packaging Poly Bottle
CAS Max % 99.53, 0.47
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Linear Formula Na2SiO3·9H2O
Concentration 1000 ppm ±10 ppm (1mL = 1mg SiO2)
Color Colorless
Solubility Soluble in water
Quantity 125 mL
Formula Weight 284.2
Grade Certified
Chemical Name or Material Silica Standard, 1000ppm (1mL = 1mg SiO2)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.