Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon carbide, 99% (metals basis)

Catalog No. p-7058828
Click to view available options
Quantity:
500 g
2 kg

CAS: 409-21-2 | CSi | 40.10 g/mol

Silicon carbide powder is used as an abrasive for such as grinding wheels, whetstone, grinding wheel and sand tiles. Silicon carbide is used to produce epitaxial grapheme by graphitization at high temperatures. It is also acts asthe metallurgical deoxidizer material. Four main application areas of SiC: functional ceramics, advanced refractory, abrasives and metallurgical materials. High purity single crystal, is used to manufacture semiconductors, manufacture of silicon carbide fibers. Silicon carbide is very hard, with excellent thermal conductivity, as a semiconductor and high temperature resistant to oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
IUPAC Name methanidylidynesilanylium
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black
Physical Form Powder
Quantity 500 g
Linear Formula SiC
Merck Index 14,8492
Solubility Information It is soluble in fused potassium hydroxide,and molten iron.It is insoluble in water (0.01mg/ml) & ethanol and acid.
Formula Weight 40.1
Percent Purity 99%
Odor Odorless
Density 3.16 g/mL
Assay (metals basis)
Chemical Name or Material Silicon carbide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 206-991-8
RTECSNumber VW0450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.