Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Silicon carbide whiskers, 99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4407786
Click to view available options
Quantity:
10 g
50 g
250 g

Abrasive For Polishing/ Grinding

Used in abrasives, polishing, grinding. It is widely applied in applications calling for high endurance, such as automobile brakes, car clutches and ceramic plates in bulletproof vests. Electronic applications of silicon carbide are the light-emitting diodes and sensors. It is utilitarian for the production of fertile, transgenic maize plants. Silicon carbide whiskers and zirconium diboride ceramic, compositely play an important role in crack-healing behavior.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 409-21-2
Molecular Formula CSi
Molecular Weight (g/mol) 40.10
MDL Number MFCD00049531
InChI Key HBMJWWWQQXIZIP-UHFFFAOYSA-N
Synonym silicon carbide, carborundum, silicon monocarbide, carborundeum, tokawhisker, betarundum, carbolon, nicalon, silundum, carbon silicide
PubChem CID 9863
ChEBI CHEBI:29390
IUPAC Name methanidylidynesilanylium
SMILES [C-]#[Si+]

Specifications

Melting Point 2700°C
Color Black to Gray
Quantity 10 g
Assay Percent Range 99% (metals basis)
Solubility Information Soluble in molten alkalis (NaOH,KOH) and molten iron. Insoluble in water.
Formula Weight 40.1
Odor Odorless
Density 3.2
Chemical Name or Material Silicon carbide whiskers
Hazard Category H350
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 206-991-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.